Learn More
Cayman Chemical LevetIracetam 10mg
SDP
Catalog No. 502894458
Encompass_Preferred
An antiepileptic agent; binds to the synaptic vesicle protein SV2A (IC50 = 1.99 μM); suppresses seizures in fully kindled rats
Catalog No. 50-289-4458 Supplier Cayman Chemical Supplier No. 900182010 MG
An antiepileptic agent; binds to the synaptic vesicle protein SV2A (IC50 = 1.99 μM); suppresses seizures in fully kindled rats