Learn More
Nova Tech LevGo smartSpatula Disposable PP Laboratory Spatula, Micro, Antistatic, Microtips, Opaque, 140 x 3.5mm (Pack of 300)
SDP
Catalog No. NC3885277
Encompass_Preferred
smartSpatulas are economical, disposable tools for transferring, collecting, and handling chemical and biological materials. Versatile double-ended design enables use as a spatula, spoon, scoop, scraper, stirring rod, or policeman. The unique hollow core minimizes thermal transfer for effective collection of liquid nitrogen frozen samples or stirring hot liquids. Virgin polypropylene construction is resistant to dilute acids and bases. Autoclavable.
Antistatic micro smartSpatula features sharp micro tips and is ideal for manipulating extremely small volumes of material. Standard spatulas feature a unique spatula/spoon end and a smaller scoop end.
The V design at the neck of the disposable laboratory spatula is a registered trademark of LevGo, Inc.
Catalog No. NC3885277 Supplier Nova Tech Supplier No. LEV17231
smartSpatulas are economical, disposable tools for transferring, collecting, and handling chemical and biological materials. Versatile double-ended design enables use as a spatula, spoon, scoop, scraper, stirring rod, or policeman. The unique hollow core minimizes thermal transfer for effective collection of liquid nitrogen frozen samples or stirring hot liquids. Virgin polypropylene construction is resistant to dilute acids and bases. Autoclavable.
Antistatic micro smartSpatula features sharp micro tips and is ideal for manipulating extremely small volumes of material. Standard spatulas feature a unique spatula/spoon end and a smaller scoop end.
The V design at the neck of the disposable laboratory spatula is a registered trademark of LevGo, Inc.