Learn More
LevitasBio LeviCell 1.0 Starter Kit, Sterile
SDP
Catalog No. 12852107
Change view
Click to view available options
For Use With (Equipment):
LeviCell 1.0 System
Catalog No. For Use With (Equipment)
12-852-107 LeviCell 1.0 System
1 options
Catalog No. 12-852-107 Supplier LevitasBio Supplier No. 1003104
Add to Cart
Add to Cart
Begin your journey into label-free cell separation and analysis with our comprehensive Starter Kit. Includes a box of 10 LeviCell sterile cartridges, reagents for 10 experiments, and everything needed for easy setup and calibration.
Begin your journey into label-free cell separation and analysis with our comprehensive Starter Kit. Includes 10 LeviCell cartridges, reagents for 10 experiments, and everything needed for easy setup and calibration.LeviCell 1.0 Starter Kit. Includes: qty 1 each of
  • LeviCell S2.3 Sterile Cartridge 10 pack
  • Levitation Agent Kit 10 Rxn
  • LeviCell 1.0 Installation and Calibration Kit
Specifications
Product Type Starter Kit
For Use With (Equipment) LeviCell 1.0 System