Learn More
LevitasBio LeviCell 1.0 Starter Kit
SDP
Catalog No. 12852106
Change view
Click to view available options
For Use With (Equipment):
LeviCell 1.0 System
Catalog No. For Use With (Equipment)
12-852-106 LeviCell 1.0 System
1 options
Catalog No. 12-852-106 Supplier LevitasBio Supplier No. 1003103
Add to Cart
Add to Cart
Begin your journey into label-free cell separation and analysis with our comprehensive Starter Kit. Includes a box of 10 LeviCell cartridges, reagents for 10 experiments, and everything needed for easy setup and calibration.
Begin your journey into label-free cell separation and analysis with our comprehensive Starter Kit. Includes 10 LeviCell cartridges, reagents for 10 experiments, and everything needed for easy setup and calibration. LeviCell 1.0 Starter Kit. Includes: qty 1 each of
  • LeviCell S2.3 Cartridge 10 pack
  • Levitation Agent Kit 10 Rxn
  • LeviCell 1.0 Installation and Calibration Kit
Specifications
Product Type Starter Kit
For Use With (Equipment) LeviCell 1.0 System