Learn More
LevitasBio LeviCell Cartridge S2.3 (Box of 10)
SDP
Catalog No. 12852110
Change view
Click to view available options
Product Type:
Cartridge
Catalog No. Product Type
12-852-110 Cartridge
1 options
Catalog No. 12-852-110 Supplier LevitasBio Supplier No. 1002010
Add to Cart
Add to Cart
The LeviCell Cartridge is a disposable cartridge designed for use with the LeviCell 1.0 system. It enables gentle, label-free separation of live cells from dead cells and debris, preserving their original state. Ideal for a wide range of applications, including oncology, neuroscience, and drug discovery.
LeviCell S2.3 Cartridge: The Heart of Gentle Cell Separation. The LeviCell S2.3 Cartridge is a disposable cartridge designed for use with the LeviCell 1.0 system. It enables gentle, label-free separation of live cells from dead cells and debris, preserving their original state.
Key Features:
  • Optically clear, inert, biocompatible plastic
  • Single-use design
  • Accommodates samples up to 350 μm in diameter
  • Ideal for a wide range of applications, including oncology, neuroscience, and drug discovery.
  • Box of 10 cartridges
Specifications
Product Type Cartridge
For Use With (Equipment) LeviCell 1.0 System