Learn More
ATCC Levilactobacillus brevis; Bb14
Supplier:  ATCC 14869
Encompass
Levilactobacillus brevis strain Bb14 is a whole-genome sequenced bacterial type strain that was isolated from feces. Growth of this strain is best in broth culture or on biphasic slants.
Catalog No. 50-238-5000