Learn More
LevitasBio LeviPrep Mouse Tissue Dissociation Kit
SDP
Catalog No. 12852103
Change view
Click to view available options
For Use With (Equipment):
LeviCell 1.0 System
Catalog No. For Use With (Equipment)
12-852-103 LeviCell 1.0 System
1 options
Catalog No. 12-852-103 Supplier LevitasBio Supplier No. 1005001
Fast and easy cold dissociation of mouse brain, lung and liver primary tissue. Combined with the LeviCell system, enrich for viable cells that are free from activation or gene expression changes. This kit allows for the gentlest handing and highest recovery preserving cellular states.
Pre-optimized kit and protocol for fast and easy cold dissociation of mouse brain, lung and liver primary tissue. Combined with the LeviCell system, enrich for the healthiest and most viable cells that are free from activation or gene expression changes. This kit allows for the gentlest handing and highest recovery preserving cellular states.
  • Includes 4 different enzymes and protocol
  • Optimized to work with the LeviCell system
  • Additional protocols available for mouse brain tissue
  • Kit Size: 24 rxns/kit
Specifications
Product Type Tissue Dissociation Kit
For Use With (Equipment) LeviCell 1.0 System