Learn More
LevitasBio LeviPrep Nuclei Kit II
SDP
Catalog No. 12852104
Change view
Click to view available options
For Use With (Equipment):
LeviCell 1.0 System
Catalog No. For Use With (Equipment)
12-852-104 LeviCell 1.0 System
1 options
Catalog No. 12-852-104 Supplier LevitasBio Supplier No. 1005055
Add to Cart
Add to Cart
Pre-optimized kit for extraction and enrichment of nuclei from fresh or frozen tissue and cells. This kit allows for gentle extraction and preservation of nuclei and clean up of debris ideal for down stream assays like single nuclei sequencing.
Pre-optimized kit for extraction and enrichment of nuclei from fresh or frozen tissue and cells. This kit allows for gentle extraction and preservation of nuclei and clean up of debris ideal for down stream assays like single nuclei sequencing.
  • Includes all reagents and plastic consumables for 16 extractions
  • Includes reagents for nuclei enrichment for use on the LeviCell systems
  • Kit Size: 16 rxns
Specifications
Product Type Nuclei Kit II
For Use With (Equipment) LeviCell 1.0 System