Learn More
LevitasBio LeviSelect Human CD45 Depletion Kit
SDP
Catalog No. 12852108
Change view
Click to view available options
Product Type:
CD45 Depletion Kit
Catalog No. Product Type
12-852-108 CD45 Depletion Kit
1 options
Catalog No. 12-852-108 Supplier LevitasBio Supplier No. 1004001
Add to Cart
Add to Cart
Depletion of human CD45+ cells from a mixture of cells with simultaneous viable cell enrichment using the LeviCell System. Human CD45+ cells are retained in the LeviCell cartridge, while the human CD45- cells of interest remain label-free and are collected for further downstream analysis.
Depletion of human CD45+ cells from a mixture of cells with simultaneous viable cell enrichment using the LeviCell System. Human CD45+ cells are retained in the LeviCell cartridge, while the human CD45- cells of interest remain label-free and are collected for further downstream analysis.
  • Includes Human anti-CD45 Nanospheres, Buffer, and Protocol
  • No centrifugation or washing steps required
  • Average 20 min. run for targeted viable cell enrichment and removal of dead, dying cell or debris
  • Kit size: 10 rxns/kit
Specifications
Product Type CD45 Depletion Kit
For Use With (Equipment) LeviCell 1.0 System