Learn More
LevitasBio LeviSelect Human CD8 T Cell Enrichment Kit
SDP
Catalog No. 12852116
Change view
Click to view available options
Product Type:
Cell Enrichment Kit
Catalog No. Product Type
12-852-116 Cell Enrichment Kit
1 options
Catalog No. 12-852-116 Supplier LevitasBio Supplier No. 1004009
Add to Cart
Add to Cart
Enrich label-free human CD8 T cells from a PBMC sample concurrently with viable cell enrichment on a LeviCell system. Viable human CD8 T cells can be used for various downstream applications. Non-CD8 T cells are magnetically labeled and remain immobilized inside the LeviCell cartridge.
Enrich label-free human CD8 T cells from a peripheral blood mononuclear cells (PBMC) sample concurrently with viable cell enrichment on a LeviCell system. Viable human CD8 T cells can be used for various downstream applications. Non-CD8 T cells are magnetically labeled and remain immobilized inside the LeviCell cartridge.
  • Includes Biotinylated-Antibody Cocktail, StrepAvidin Nanospheres, Buffer, and Protocol
  • No centrifugation or washing steps required
  • Average 20 min. run for targeted viable cell enrichment and removal of dead, dying cell or debris
  • Kit size: 10 rxns / kit
Specifications
Product Type Cell Enrichment Kit
For Use With (Equipment) LeviCell 1.0 System