Learn More
LevitasBio LeviSelect Mouse Tissue RBC Depletion Kit
SDP
Catalog No. 12852101
Change view
Click to view available options
For Use With (Equipment):
LeviCell 1.0 System
Catalog No. For Use With (Equipment)
12-852-101 LeviCell 1.0 System
1 options
Catalog No. 12-852-101 Supplier LevitasBio Supplier No. 1004110
Add to Cart
Add to Cart
The LeviSelect Mouse Tissue RBC Depletion Kit has been designed to deplete up to 5 million mature erythrocyte and erythroid precursor cells per reaction after a mechanical or enzymatic tissue dissociation while simultaneously enriching for live cells.
The LeviSelect Mouse Tissue RBC Depletion Kit has been designed to deplete up to 5 million mature erythrocyte and erythroid precursor cells per reaction after a mechanical or enzymatic tissue dissociation while simultaneously enriching for live cells.
  • Includes Mouse RBC Ab Cocktail, SAV Nanospheres, buffer and protocol
  • Average 20 min. run for removal of RBCs and enrichment of viable cells away from debris and dead cells
  • Kit size: 10 rxns / kit
Specifications
Product Type RBC Depletion Kit
For Use With (Equipment) LeviCell 1.0 System