Learn More
Cayman Chemical LevIstolIde A 10mg
SDP
Catalog No. 502949220
Encompass_Preferred
A phthalide with diverse biological activities; inhibits TPA and A23187-induced histamine release from HMC-1 cells at 10 µM; induces the production of ROS, ER stress, and apoptosis in HCT116 cells at 100 µM; prevents ethanol-induced gastric ulcers in rats at 10 mg/kg; decreases aggregation of amyloid-β in brain and improves spatial and learning memory in the APP/PS1 transgenic mouse model of Alzheimer's disease at 2 mg/kg
Catalog No. 50-294-9220 Supplier Cayman Chemical Supplier No. 3438610 MG
A phthalide with diverse biological activities; inhibits TPA and A23187-induced histamine release from HMC-1 cells at 10 µM; induces the production of ROS, ER stress, and apoptosis in HCT116 cells at 100 µM; prevents ethanol-induced gastric ulcers in rats at 10 mg/kg; decreases aggregation of amyloid-β in brain and improves spatial and learning memory in the APP/PS1 transgenic mouse model of Alzheimer's disease at 2 mg/kg