Learn More
LevitasBio Levitation Agent
SDP
Catalog No. 12852112
Change view
Click to view available options
For Use With (Equipment):
LeviCell 1.0 System
Catalog No. For Use With (Equipment)
12-852-112 LeviCell 1.0 System
1 options
Catalog No. 12-852-112 Supplier LevitasBio Supplier No. 1003001
Add to Cart
Add to Cart
A key component of the LeviCell system, this levitation agent enables gentle, label-free cell separation. It interacts with cells based on membrane integrity, allowing for the isolation of viable cells while leaving dead cells and debris behind.
A key component of the LeviCell system, this levitation agent enables gentle, label-free cell separation. It interacts with cells based on membrane integrity, allowing for the isolation of viable cells while leaving dead cells and debris behind.
Specifications
Product Type Levitation Agent
For Use With (Equipment) LeviCell 1.0 System