Learn More
Apexbio Technology LLC Levobetaxolol HCl 116209-55-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000094121
Encompass_Preferred
Levobetaxolol HCl (CAS 116209-55-3) is a selective -adrenergic receptor antagonist It demonstrates a high binding affinity for human 1-adrenergic receptors (K sub i /sub 0 76 nM) and lower affinity for 2-adrenergic receptors (K sub i /sub 32 6 nM) indicating pronounced selectivity toward the 1 subtype Through inhibition of -adrenergic signaling Levobetaxolol HCl is utilized in research investigating cardiovascular regulation receptor pharmacology and the molecular mechanisms underlying 1-adrenergic blockade Its well-characterized receptor selectivity supports its use in dissecting -adrenergic pathway contributions in preclinical models
Catalog No. 50-000-94121 Supplier Apexbio Technology LLC Supplier No. B17785.1
Levobetaxolol HCl (CAS 116209-55-3) is a selective -adrenergic receptor antagonist It demonstrates a high binding affinity for human 1-adrenergic receptors (K sub i /sub 0 76 nM) and lower affinity for 2-adrenergic receptors (K sub i /sub 32 6 nM) indicating pronounced selectivity toward the 1 subtype Through inhibition of -adrenergic signaling Levobetaxolol HCl is utilized in research investigating cardiovascular regulation receptor pharmacology and the molecular mechanisms underlying 1-adrenergic blockade Its well-characterized receptor selectivity supports its use in dissecting -adrenergic pathway contributions in preclinical models