Learn More
TARGETMOL CHEMICALS INC LEVOBETAXOLOL HYDROCHLOR 100MG
SDP
Catalog No. 502576491
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Levobetaxolol hydrochloride ((S)-Betaxolol hydrochloride) is a beta-adrenergic receptor inhibitor (beta blocker) used to lower the pressure in the eye in treating conditions such as glaucoma. purity: 99%
Catalog No. 50-257-6491 Supplier TARGETMOL CHEMICALS INC Supplier No. T6565100MG
Also available in 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Levobetaxolol hydrochloride ((S)-Betaxolol hydrochloride) is a beta-adrenergic receptor inhibitor (beta blocker) used to lower the pressure in the eye in treating conditions such as glaucoma. purity: 99%