Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Levobetaxolol hydrochloride | 116209-55-3 | 99.3% | 1 ML
SDP

Catalog No. 5000407161
Encompass_Preferred

Levobetaxolol hydrochloride is a beta-adrenergic receptor inhibitor (beta blocker) that effectively reduces pressure within the eye. This mechanism allows it to be suitable for research related to glaucoma. It presents as a solid, with a color ranging from white to off-white.

  • Functions as a beta-adrenergic receptor inhibitor (beta blocker).
  • Available as a solid, appearing white to off-white.
  • Purity: 99.3%.
  • CAS number: 116209-55-3.
  • Molecular weight: 343.89.
  • Formula: C18H30ClNO3.
  • Applicable for research purposes, particularly in the study of glaucoma.

Catalog No. 50-004-07161 Supplier Medchemexpress LLC Supplier No. HYB0381B10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Levobetaxolol hydrochloride is a beta-adrenergic receptor inhibitor (beta blocker) that effectively reduces pressure within the eye. This mechanism allows it to be suitable for research related to glaucoma. It presents as a solid, with a color ranging from white to off-white.

  • Functions as a beta-adrenergic receptor inhibitor (beta blocker).
  • Available as a solid, appearing white to off-white.
  • Purity: 99.3%.
  • CAS number: 116209-55-3.
  • Molecular weight: 343.89.
  • Formula: C18H30ClNO3.
  • Applicable for research purposes, particularly in the study of glaucoma.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.