Learn More
Medchemexpress LLC Levobetaxolol hydrochloride | 116209-55-3 | 99.31% | 50 MG
SDP
Catalog No. 5000405430
Encompass_Preferred
Levobetaxolol hydrochloride is a beta-adrenergic receptor inhibitor, commonly known as a beta blocker, which has the ability to reduce pressure within the eye. This compound is utilized in the research of glaucoma.
  • Beta-adrenergic receptor inhibitor (beta blocker)
  • Capable of lowering intraocular pressure
  • Applicable for glaucoma research
Catalog No. 50-004-05430 Supplier Medchemexpress LLC Supplier No. HYB0381B50MG
Levobetaxolol hydrochloride is a beta-adrenergic receptor inhibitor, commonly known as a beta blocker, which has the ability to reduce pressure within the eye. This compound is utilized in the research of glaucoma.
  • Beta-adrenergic receptor inhibitor (beta blocker)
  • Capable of lowering intraocular pressure
  • Applicable for glaucoma research