Learn More
Cayman Chemical Levobetaxolol (hydrochloride), Purity: ≥98%, CAS: 116209-55-3, 50 mg
SDP
Catalog No. 502949206
Encompass_Preferred
A β1-adrenergic receptor antagonist (Ki = 0.76 nM for the human receptor); selective for β1-ARs over β2-ARs (Ki = 32.6 nM), as well as a panel of 89 additional receptors (IC50s = >1 µM); inhibits isoproterenol-induced cAMP production in human non-pigmented ciliary epithelial cells (Ki = 16.4 nM); topical application reduces intraocular pressure in a cynomolgus monkey model of ocular hypertension at 150 µg/eye
Catalog No. 50-294-9206 Supplier Cayman Chemical Supplier No. 3343550 MG
A β1-adrenergic receptor antagonist (Ki = 0.76 nM for the human receptor); selective for β1-ARs over β2-ARs (Ki = 32.6 nM), as well as a panel of 89 additional receptors (IC50s = >1 µM); inhibits isoproterenol-induced cAMP production in human non-pigmented ciliary epithelial cells (Ki = 16.4 nM); topical application reduces intraocular pressure in a cynomolgus monkey model of ocular hypertension at 150 µg/eye