Learn More
Cayman Chemical LevobunololhydrochlorIde 100mg
SDP
Catalog No. 502981566
Encompass_Preferred
A β-adrenergic receptor antagonist that acts as a vasodilator and increases ocular circulation; formulations containing levobunolol are used to manage glaucoma
Catalog No. 50-298-1566 Supplier Cayman Chemical Supplier No. 21005100 MG
A β-adrenergic receptor antagonist that acts as a vasodilator and increases ocular circulation; formulations containing levobunolol are used to manage glaucoma