Learn More
Cayman Chemical Levobunolol (hydrochloride), Purity: ≥98%, CAS: 27912-14-7, 50 mg
SDP
Catalog No. 502982757
Encompass_Preferred
A β-adrenergic receptor antagonist that acts as a vasodilator and increases ocular circulation; formulations containing levobunolol are used to manage glaucoma
Catalog No. 50-298-2757 Supplier Cayman Chemical Supplier No. 2100550 MG
A β-adrenergic receptor antagonist that acts as a vasodilator and increases ocular circulation; formulations containing levobunolol are used to manage glaucoma