Learn More
Selleck Chemical LLC Levobupivacaine S5181-25mg
SDP
Catalog No. 503112346
Encompass_Preferred
Levobupivacaine (Chirocaine (S)-(-)-Bupivacaine L-(-)-Bupivacaine) the S (-)-enantiomer of bupivacaine is a long-acting amide local anaesthetic used in analgesia and anaesthesia Levobupivacaine exhibits effects on motor and sensory nerves by inhibiting the opening of voltage-gated sodium channels
Catalog No. 50-311-2346 Supplier Selleck Chemical LLC Supplier No. S518125MG
Levobupivacaine (Chirocaine (S)-(-)-Bupivacaine L-(-)-Bupivacaine) the S (-)-enantiomer of bupivacaine is a long-acting amide local anaesthetic used in analgesia and anaesthesia Levobupivacaine exhibits effects on motor and sensory nerves by inhibiting the opening of voltage-gated sodium channels