Learn More
Apexbio Technology LLC Levobupivacaine HCl 27262-48-2 50mg
SDP
Catalog No. 5000096489
Encompass_Preferred
Levobupivacaine HCl (CAS 27262-48-2) is the optically pure S(-)-enantiomer of bupivacaine and functions as a long-acting local anesthetic It exerts its effects by reversibly blocking voltage-gated sodium channels in neuronal cell membranes thereby inhibiting the initiation and propagation of action potentials This action results in a localized loss of sensory and motor function Levobupivacaine HCl is widely utilized in biomedical research to study nociceptive pathways investigate anesthetic pharmacology and evaluate neurotoxicity or cardiotoxicity profiles of local anesthetics in both in vitro and in vivo models
Catalog No. 50-000-96489 Supplier Apexbio Technology LLC Supplier No. B178450
Levobupivacaine HCl (CAS 27262-48-2) is the optically pure S(-)-enantiomer of bupivacaine and functions as a long-acting local anesthetic It exerts its effects by reversibly blocking voltage-gated sodium channels in neuronal cell membranes thereby inhibiting the initiation and propagation of action potentials This action results in a localized loss of sensory and motor function Levobupivacaine HCl is widely utilized in biomedical research to study nociceptive pathways investigate anesthetic pharmacology and evaluate neurotoxicity or cardiotoxicity profiles of local anesthetics in both in vitro and in vivo models