Learn More
Selleck Chemical LLC Levobupivacaine HCl S4061-50mg
SDP
Catalog No. 503109923
Encompass_Preferred
Levobupivacaine HCl ((S)-(-)-Bupivacaine) the pure S(-)-enantiomer of bupivacaine is a reversible neuronal sodium channel inhibitor used as a long-acting local anesthetic
Catalog No. 50-310-9923 Supplier Selleck Chemical LLC Supplier No. S406150MG
Levobupivacaine HCl ((S)-(-)-Bupivacaine) the pure S(-)-enantiomer of bupivacaine is a reversible neuronal sodium channel inhibitor used as a long-acting local anesthetic