Learn More
Cayman Chemical Levobupivacaine (hydrochloride), Purity: ≥98%, CAS: 27262-48-2, 100 mg
SDP
Catalog No. 502992043
Encompass_Preferred
A sodium channel blocker and local anesthetic; levorotary stereoisomer of bupivacaine; inhibits INa by 57.8% in isolated guinea pig ventricular myocytes at 10 μM; reduces the Vmax and action potential duration to 76.7 and 83.4% of controls, respectively, in isolated guinea pig papillary muscle at 10 μM; induces a complete block of proprioception, motor function, and nociception in the hindleg of rats at 7.7 mM by percutaneous injection into the sciatic nerve
Catalog No. 50-299-2043 Supplier Cayman Chemical Supplier No. 31596100 MG
A sodium channel blocker and local anesthetic; levorotary stereoisomer of bupivacaine; inhibits INa by 57.8% in isolated guinea pig ventricular myocytes at 10 μM; reduces the Vmax and action potential duration to 76.7 and 83.4% of controls, respectively, in isolated guinea pig papillary muscle at 10 μM; induces a complete block of proprioception, motor function, and nociception in the hindleg of rats at 7.7 mM by percutaneous injection into the sciatic nerve