Learn More
Medchemexpress LLC Levobupivacaine hydrochloride | 27262-48-2 | >99.8% | 5 MG
SDP
Catalog No. 5000408521
Encompass_Preferred
Levobupivacaine hydrochloride is a long-acting amide local agent that can suppress or relieve pain. It exerts analgesic effects through reversible blockade of neuronal sodium channels. It can inhibit impulse transmission and conduction in cardiovascular and other tissues, possessing certain cardiac and CNS toxicity. It is metabolized by hepatic cytochrome P450 (CYP450) enzymes in vivo and can also induce ferroptosis by miR-489-3p/SLC7A11 signaling in gastric cancer.
  • Long-acting amide local agent
  • Suppresses or relieves pain
  • Exerts analgesic effects through reversible blockade of neuronal sodium channels
  • Inhibits impulse transmission and conduction in cardiovascular and other tissues
  • Metabolized by hepatic cytochrome P450 (CYP450) enzymes in vivo
  • Induces ferroptosis by miR-489-3p/SLC7A11 signaling in gastric cancer
Catalog No. 50-004-08521 Supplier Medchemexpress LLC Supplier No. HYB0653A5MG
Levobupivacaine hydrochloride is a long-acting amide local agent that can suppress or relieve pain. It exerts analgesic effects through reversible blockade of neuronal sodium channels. It can inhibit impulse transmission and conduction in cardiovascular and other tissues, possessing certain cardiac and CNS toxicity. It is metabolized by hepatic cytochrome P450 (CYP450) enzymes in vivo and can also induce ferroptosis by miR-489-3p/SLC7A11 signaling in gastric cancer.
  • Long-acting amide local agent
  • Suppresses or relieves pain
  • Exerts analgesic effects through reversible blockade of neuronal sodium channels
  • Inhibits impulse transmission and conduction in cardiovascular and other tissues
  • Metabolized by hepatic cytochrome P450 (CYP450) enzymes in vivo
  • Induces ferroptosis by miR-489-3p/SLC7A11 signaling in gastric cancer