Learn More
Medchemexpress LLC Levobupivacaine hydrochloride | 27262-48-2 | 99.86% | 100 MG
SDP
Catalog No. 5000408441
Encompass_Preferred
Levobupivacaine hydrochloride is a long-acting amide local agent designed to suppress or relieve pain. It functions by reversibly blocking neuronal sodium channels, inhibiting impulse transmission and conduction. This agent is metabolized by hepatic cytochrome P450 enzymes and can induce ferroptosis through miR-489-3p/SLC7A11 signaling in gastric cancer. It exhibits certain cardiac and CNS toxicity and is intended for research use only.
  • Long-acting amide local agent
  • Suppresses or relieves pain
  • Reversibly blocks neuronal sodium channels
  • Inhibits impulse transmission and conduction
  • Metabolized by hepatic cytochrome P450 enzymes
  • Induces ferroptosis by miR-489-3p/SLC7A11 signaling
  • Targets sodium channels and ferroptosis
Catalog No. 50-004-08441 Supplier Medchemexpress LLC Supplier No. HYB0653A100MG
Add to Cart
Add to Cart
Levobupivacaine hydrochloride is a long-acting amide local agent designed to suppress or relieve pain. It functions by reversibly blocking neuronal sodium channels, inhibiting impulse transmission and conduction. This agent is metabolized by hepatic cytochrome P450 enzymes and can induce ferroptosis through miR-489-3p/SLC7A11 signaling in gastric cancer. It exhibits certain cardiac and CNS toxicity and is intended for research use only.
  • Long-acting amide local agent
  • Suppresses or relieves pain
  • Reversibly blocks neuronal sodium channels
  • Inhibits impulse transmission and conduction
  • Metabolized by hepatic cytochrome P450 enzymes
  • Induces ferroptosis by miR-489-3p/SLC7A11 signaling
  • Targets sodium channels and ferroptosis