Learn More
Medchemexpress LLC Levobupivacaine hydrochloride | 27262-48-2 | 99.9% | 200 MG
SDP
Catalog No. 5000407716
Encompass_Preferred
Levobupivacaine hydrochloride is a long-acting amide local agent primarily used for pain management. It functions by reversibly blocking neuronal sodium channels, which inhibits impulse transmission in various tissues, including cardiovascular. This compound is metabolized by hepatic cytochrome P450 enzymes and can also induce ferroptosis through miR-489-3p/SLC7A11 signaling in gastric cancer. It is intended for research use only.
  • Long-acting amide local agent
  • Suppresses or relieves pain
  • Reversibly blocks neuronal sodium channels
  • Inhibits impulse transmission and conduction in cardiovascular and other tissues
  • Metabolized by hepatic cytochrome P450 enzymes
  • Can induce ferroptosis by miR-489-3p/SLC7A11 signaling in gastric cancer
  • For research use only
Catalog No. 50-004-07716 Supplier Medchemexpress LLC Supplier No. HYB0653A200MG
Add to Cart
Add to Cart
Levobupivacaine hydrochloride is a long-acting amide local agent primarily used for pain management. It functions by reversibly blocking neuronal sodium channels, which inhibits impulse transmission in various tissues, including cardiovascular. This compound is metabolized by hepatic cytochrome P450 enzymes and can also induce ferroptosis through miR-489-3p/SLC7A11 signaling in gastric cancer. It is intended for research use only.
  • Long-acting amide local agent
  • Suppresses or relieves pain
  • Reversibly blocks neuronal sodium channels
  • Inhibits impulse transmission and conduction in cardiovascular and other tissues
  • Metabolized by hepatic cytochrome P450 enzymes
  • Can induce ferroptosis by miR-489-3p/SLC7A11 signaling in gastric cancer
  • For research use only