Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Levobupivacaine hydrochloride, 98%

Catalog No. AAH3764303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH37643-03 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH37643-03 Supplier Thermo Scientific™ Supplier No. H3764303
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 27262-48-2
Molecular Formula C18H29ClN2O
MDL Number MFCD01704265
Quantity 1 g
InChI Key SIEYLFHKZGLBNX-NTISSMGPSA-N
SMILES [H+].[Cl-].CCCCN1CCCC[C@H]1C(=O)NC1=C(C)C=CC=C1C
IUPAC Name hydrogen (2S)-1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide chloride
Molecular Weight (g/mol) 324.89
Formula Weight 324.89
Percent Purity 98%
Physical Form Solid
Show More Show Less
ShelfLife 5 Years
EINECSNumber 642-955-1
RTECSNumber TK6120000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.