Learn More
Medchemexpress LLC Levocarnitine propionate-d9 hydrochloride | 2687961-04-0 | 99.0% | 1 MG
SDP
Catalog No. 5000408056
Encompass_Preferred
Levocarnitine propionate-d9 hydrochloride is a deuterium-labeled form of Levocarnitine propionate hydrochloride. This compound is used in studies related to renal function deterioration, congestive heart failure, and intermittent claudication. It can serve as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Deuterium-labeled form of Levocarnitine propionate hydrochloride
  • Useful in studies related to renal function deterioration, congestive heart failure, and intermittent claudication
  • Can serve as a tracer
  • Can be used as an internal standard for quantitative analysis
  • For research use only
Catalog No. 50-004-08056 Supplier Medchemexpress LLC Supplier No. HYB0932S21MG
Levocarnitine propionate-d9 hydrochloride is a deuterium-labeled form of Levocarnitine propionate hydrochloride. This compound is used in studies related to renal function deterioration, congestive heart failure, and intermittent claudication. It can serve as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Deuterium-labeled form of Levocarnitine propionate hydrochloride
  • Useful in studies related to renal function deterioration, congestive heart failure, and intermittent claudication
  • Can serve as a tracer
  • Can be used as an internal standard for quantitative analysis
  • For research use only