Learn More
Medchemexpress LLC Levocarnitine propionate (hydrochloride) | 119793-66-7 | 95.0% | 25 MG

Supplier: Medchemexpress LLC 25MGHYB0932
Levocarnitine propionate hydrochloride is a chemical compound used in the treatment of various conditions such as renal function deterioration, congestive heart failure, and intermittent claudication. It is intended for research use only.
- Molecular formula: C10H20ClNO4
- Molecular weight: 253.72
- Appears as a white to off-white solid
- Melting point of 165-167 °C
- Stable under recommended storage conditions
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.