Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Levocarnitine propionate hydrochloride | 119793-66-7 | 95.0% | 500 MG
SDP

Supplier:  Medchemexpress LLC HYB0932500MG

Encompass_Preferred

Levocarnitine propionate hydrochloride (L-Propionylcarnitine chloride) is utilized for addressing the deterioration of renal function, congestive heart failure, intermittent claudication, and various other diseases. This product is for research use only and not sold to patients.

  • Molecular weight: 253.72
  • Chemical formula: C10H20ClNO4
  • Appearance: Solid
  • Color: White to off-white
  • Ships at room temperature in continental US
  • Store solid at 4°C, sealed, away from moisture
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month, sealed, away from moisture
  • Soluble in DMSO at 250 mg/mL (requires ultrasonic, use newly opened DMSO)
  • Soluble in H2O at 60 mg/mL (requires ultrasonic)

Catalog No. 50-004-09090


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.