Learn More
AdipoGen Levocetirizine dihydrochloride (10 mg)
SDP
Supplier:  AdipoGen CDXL0121M010
Encompass_Preferred
Chemical. CAS: 130018-87-0. Formula: C21H25ClN2O3 . 2HCl. MW: 461.81. Levocetirizine hydrochloride is the (R)-enantiomer of the histamine H1 receptor antagonist and antihistamine agent cetirizine. It binds to the H1 receptor with higher affinity than cetirizine and is 25,000-100,000-fold selective for H1 receptors over muscarinic M1-M5 receptors. Intranasal administration dose-dependently decreases histamine-induced nasal rubbing and sneezing in mice. Has been used in the treatment of allergic rhinitis and chronic idiopathic urticaria.
Catalog No. 50-205-4523