Learn More
Apexbio Technology LLC Levocetirizine-DiHCl
SDP
Catalog No. 5000568741
Encompass_Preferred
Levocetirizine Dihydrochloride (CAS No 130018-87-0) is an antihistamine used in biomedical research particularly in allergy and immunology studies It is a third-generation non-sedative H1 receptor antagonist primarily derived from the metabolic process of cetirizine Its mechanism of action involves selective inhibition of the peripheral histamine H1 receptors thereby blocking the pathway responsible for allergic symptoms The compound exhibits in vitro activity in the nanomolar to low micromolar range Levocetirizine is commonly applied in various cell models and animal studies to evaluate its efficacy in reducing inflammatory responses and allergic reactions It is also utilized in drug screening processes due to its well-documented pharmacological profile Experimental concentrations typically depend on the specific research design employed ensuring the desired therapeutic or investigative outcomes
Catalog No. 50-005-68741 Supplier Apexbio Technology LLC Supplier No. C648325
Levocetirizine Dihydrochloride (CAS No 130018-87-0) is an antihistamine used in biomedical research particularly in allergy and immunology studies It is a third-generation non-sedative H1 receptor antagonist primarily derived from the metabolic process of cetirizine Its mechanism of action involves selective inhibition of the peripheral histamine H1 receptors thereby blocking the pathway responsible for allergic symptoms The compound exhibits in vitro activity in the nanomolar to low micromolar range Levocetirizine is commonly applied in various cell models and animal studies to evaluate its efficacy in reducing inflammatory responses and allergic reactions It is also utilized in drug screening processes due to its well-documented pharmacological profile Experimental concentrations typically depend on the specific research design employed ensuring the desired therapeutic or investigative outcomes