Learn More
Medchemexpress LLC Levocetirizine dihydrochloride | 130018-87-0 | 99.8% | 461.81 | 100 MG
SDP
Catalog No. 5000452502
Encompass_Preferred
Levocetirizine dihydrochloride is a third-generation peripheral H1-receptor antagonist. It is the R-enantiomer of Cetirizine and acts as an antihistaminic agent. This compound exhibits a higher affinity for the histamine H1-receptor compared to (S)-Cetirizine and is effective in treating allergic rhinitis and chronic idiopathic urticaria.
  • Third-generation peripheral H1-receptor antagonist
  • R-enantiomer of Cetirizine
  • Exhibits higher affinity for histamine H1-receptor
  • Effective in treating allergic rhinitis
  • Used for chronic idiopathic urticaria
Catalog No. 50-004-52502 Supplier Medchemexpress LLC Supplier No. HYW010841100MG
Add to Cart
Add to Cart
Levocetirizine dihydrochloride is a third-generation peripheral H1-receptor antagonist. It is the R-enantiomer of Cetirizine and acts as an antihistaminic agent. This compound exhibits a higher affinity for the histamine H1-receptor compared to (S)-Cetirizine and is effective in treating allergic rhinitis and chronic idiopathic urticaria.
  • Third-generation peripheral H1-receptor antagonist
  • R-enantiomer of Cetirizine
  • Exhibits higher affinity for histamine H1-receptor
  • Effective in treating allergic rhinitis
  • Used for chronic idiopathic urticaria