Learn More
Medchemexpress LLC Levocetirizine dihydrochloride | 130018-87-0 | 99.8% | 461.81 | 50 MG
SDP
Catalog No. 5000367024
Encompass_Preferred
Levocetirizine dihydrochloride is a third-generation peripheral H1-receptor antagonist. It is an antihistaminic agent which is the R-enantiomer of Cetirizine. This compound has a higher affinity for the histamine H1-receptor than (S)-Cetirizine and can effectively treat allergic rhinitis and chronic idiopathic urticaria.
  • Third-generation peripheral H1-receptor antagonist
  • R-enantiomer of Cetirizine
  • Higher affinity for histamine H1-receptor
  • Effective for allergic rhinitis and chronic idiopathic urticaria
Catalog No. 50-003-67024 Supplier Medchemexpress LLC Supplier No. HYW01084150MG
Levocetirizine dihydrochloride is a third-generation peripheral H1-receptor antagonist. It is an antihistaminic agent which is the R-enantiomer of Cetirizine. This compound has a higher affinity for the histamine H1-receptor than (S)-Cetirizine and can effectively treat allergic rhinitis and chronic idiopathic urticaria.
  • Third-generation peripheral H1-receptor antagonist
  • R-enantiomer of Cetirizine
  • Higher affinity for histamine H1-receptor
  • Effective for allergic rhinitis and chronic idiopathic urticaria