Learn More
Selleck Chemical LLC Levocetirizine Dihydrochloride S4849-25mg
SDP
Catalog No. 503108984
Encompass_Preferred
Levocetirizine (Xyzal (-)-Cetirizine) is the active R-enantiomer of cetirizine and represents a new second-generation histamine H1 antagonist
Catalog No. 50-310-8984 Supplier Selleck Chemical LLC Supplier No. S484925MG
Levocetirizine (Xyzal (-)-Cetirizine) is the active R-enantiomer of cetirizine and represents a new second-generation histamine H1 antagonist