Learn More
Apexbio Technology LLC Levodopa 59-92-7 1g
SDP
Catalog No. 5000095244
Encompass_Preferred
Levodopa (59-92-7) is a small-molecule precursor of dopamine It modulates dopaminergic signaling pathways thereby influencing neurotransmission and associated neuronal functions Levodopa exerts its biological activity primarily through enzymatic decarboxylation by aromatic L-amino acid decarboxylase within dopaminergic neurons yielding dopamine In in vitro assays levodopa demonstrates cholinergic receptor antagonism with reported IC50 values typically ranging 10 30 M depending on specific cellular and experimental conditions Based on these pharmacological properties levodopa holds research potential in studies of dopaminergic signaling neuronal plasticity motor function modulation preclinical Parkinson s disease models and stroke recovery investigations
Catalog No. 50-000-95244 Supplier Apexbio Technology LLC Supplier No. A84601000
Levodopa (59-92-7) is a small-molecule precursor of dopamine It modulates dopaminergic signaling pathways thereby influencing neurotransmission and associated neuronal functions Levodopa exerts its biological activity primarily through enzymatic decarboxylation by aromatic L-amino acid decarboxylase within dopaminergic neurons yielding dopamine In in vitro assays levodopa demonstrates cholinergic receptor antagonism with reported IC50 values typically ranging 10 30 M depending on specific cellular and experimental conditions Based on these pharmacological properties levodopa holds research potential in studies of dopaminergic signaling neuronal plasticity motor function modulation preclinical Parkinson s disease models and stroke recovery investigations