Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Levodopa Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012221 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2221 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2221 Supplier MilliporeSigma Supelco Supplier No. PHR1271500MG
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 59-92-7
Physical Form Neat
Linear Formula (HO)2C6H3 CH2 CH(NH2)CO2 H
MDL Number MFCD00002598
Melting Point 276°C to 278°C
Synonym 3,4-Dihydroxy-L-phenylalanine; 3-(3,4-Dihydroxyphenyl)-L-alanine; L-3-Hydroxytyrosine; L-DOPA; Levodopa
Type Certified Reference Material
Quantity 500 mg
Formula Weight 197.19
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.