Learn More
Medchemexpress LLC Levodropropizine | 99291-25-5 | MFCD00866852 | 99.7% | 10 G
SDP
Catalog No. 5000409796
Encompass_Preferred
Levodropropizine (DF-526) is an orally active histamine receptor inhibitor, described as an effective and very well tolerated peripheral antitussive agent. It also suppresses seizure activity and reduces plasma extravasation in rat models.
  • Orally active histamine receptor inhibitor.
  • Effective peripheral antitussive agent.
  • Well tolerated.
  • Inhibits seizure activity.
  • Reduces plasma extravasation.
  • Treats cough in adults and children.
  • Attenuates chronic cigarette smoke-exposure enhanced cough in guinea pigs.
  • Suppresses seizure activity in rats with pentylenetetrazol-induced epilepsy.
Catalog No. 50-004-09796 Supplier Medchemexpress LLC Supplier No. HYB189510G
Add to Cart
Add to Cart
Levodropropizine (DF-526) is an orally active histamine receptor inhibitor, described as an effective and very well tolerated peripheral antitussive agent. It also suppresses seizure activity and reduces plasma extravasation in rat models.
  • Orally active histamine receptor inhibitor.
  • Effective peripheral antitussive agent.
  • Well tolerated.
  • Inhibits seizure activity.
  • Reduces plasma extravasation.
  • Treats cough in adults and children.
  • Attenuates chronic cigarette smoke-exposure enhanced cough in guinea pigs.
  • Suppresses seizure activity in rats with pentylenetetrazol-induced epilepsy.