Learn More
Medchemexpress LLC Levofloxacin-13C,d3 | 1261398-33-7 | 98.8% | 1 MG
SDP
Catalog No. 5000406679
Encompass_Preferred
Levofloxacin-13C,d3 is a labeled version of Levofloxacin, an antibiotic commonly used to treat a variety of bacterial infections. This specific compound incorporates 13C and deuterium isotopes, making it suitable for research applications where isotopically labeled compounds are required, such as in pharmacokinetic studies or as an internal standard in analytical methods.
  • Isotopically labeled for research applications
  • Suitable for pharmacokinetic studies
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06679 Supplier Medchemexpress LLC Supplier No. HYB0330S21MG
Levofloxacin-13C,d3 is a labeled version of Levofloxacin, an antibiotic commonly used to treat a variety of bacterial infections. This specific compound incorporates 13C and deuterium isotopes, making it suitable for research applications where isotopically labeled compounds are required, such as in pharmacokinetic studies or as an internal standard in analytical methods.
  • Isotopically labeled for research applications
  • Suitable for pharmacokinetic studies
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS