Learn More
Apexbio Technology LLC Levofloxacin 100986-85-4 5g
SDP
Catalog No. 502240332
Encompass_Preferred
Levofloxacin (CAS 100986-85-4) is a small-molecule inhibitor targeting bacterial DNA gyrase (topoisomerase II) It modulates DNA supercoiling thereby regulating DNA replication and transcription in bacteria Levofloxacin exerts its biological activity primarily through stabilization of the DNA enzyme cleavage intermediate interfering with DNA replication and causing bacterial cell death In in vitro studies Levofloxacin demonstrates antibacterial inhibitory activity with IC50 values typically ranging from approximately 0 5 g/mL to 4 g/mL depending on bacterial species and experimental conditions Based on these pharmacological properties Levofloxacin holds research potential in studies involving bacterial susceptibility profiling and the development of antibiotic resistance
This item is not returnable. View return policy
Catalog No. 50-224-0332 Supplier Apexbio Technology LLC Supplier No. B19595000
Levofloxacin (CAS 100986-85-4) is a small-molecule inhibitor targeting bacterial DNA gyrase (topoisomerase II) It modulates DNA supercoiling thereby regulating DNA replication and transcription in bacteria Levofloxacin exerts its biological activity primarily through stabilization of the DNA enzyme cleavage intermediate interfering with DNA replication and causing bacterial cell death In in vitro studies Levofloxacin demonstrates antibacterial inhibitory activity with IC50 values typically ranging from approximately 0 5 g/mL to 4 g/mL depending on bacterial species and experimental conditions Based on these pharmacological properties Levofloxacin holds research potential in studies involving bacterial susceptibility profiling and the development of antibiotic resistance