Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Levofloxacin, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012283 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2283 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2283 Supplier MilliporeSigma Supelco Supplier No. 40922100MG
Add to Cart
Add to Cart

Chemical structure: quinolone

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 100986-85-4
Physical Form Neat
Linear Formula C18H20 FN3O4
MDL Number MFCD00865049
Synonym (-)-Ofloxacin
Type Analytical Standard
Quantity 100 mg
Formula Weight 361.37
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.