Learn More
Medchemexpress LLC Levofloxacin-d8 | 1217716-71-6 | 99.8% | 5 MG
SDP
Catalog No. 5000406376
Encompass_Preferred
Levofloxacin-d8 is the deuterium-labeled version of Levofloxacin. Levofloxacin is a synthetic fluoroquinolone and an antibacterial agent that inhibits the supercoiling activity of bacterial DNA gyrase, thereby halting DNA replication. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06376 Supplier Medchemexpress LLC Supplier No. HYB0330S5MG
Levofloxacin-d8 is the deuterium-labeled version of Levofloxacin. Levofloxacin is a synthetic fluoroquinolone and an antibacterial agent that inhibits the supercoiling activity of bacterial DNA gyrase, thereby halting DNA replication. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS