Learn More
Medchemexpress LLC Levofloxacin-d8 hydrochloride | 2699607-50-4 | 99.0% | 1 MG
SDP
Catalog No. 5000406359
Encompass_Preferred
Levofloxacin-d8 hydrochloride is a deuterated form of levofloxacin, an antibiotic from the fluoroquinolone family. This product is suitable for research applications requiring a stable-labeled internal standard or for studying drug metabolism.
  • High purity for reliable results
  • Stable under recommended storage conditions
  • Suitable for laboratory use
Catalog No. 50-004-06359 Supplier Medchemexpress LLC Supplier No. HYB0330BS1MG
Levofloxacin-d8 hydrochloride is a deuterated form of levofloxacin, an antibiotic from the fluoroquinolone family. This product is suitable for research applications requiring a stable-labeled internal standard or for studying drug metabolism.
  • High purity for reliable results
  • Stable under recommended storage conditions
  • Suitable for laboratory use