Learn More
Medchemexpress LLC Levofloxacin-d8 ((-)-ofloxacin-d8) | 1217716-71-6 | 99.8% | 1 MG
SDP
Catalog No. 5000405987
Encompass_Preferred
Levofloxacin-d8 is a deuterium-labeled version of Levofloxacin. Levofloxacin is a synthetic fluoroquinolone that acts as an antibacterial agent by inhibiting the supercoiling activity of bacterial DNA gyrase, thereby halting DNA replication.
  • Deuterium labeled levofloxacin
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only
Catalog No. 50-004-05987 Supplier Medchemexpress LLC Supplier No. HYB0330S1MG
Add to Cart
Add to Cart
Levofloxacin-d8 is a deuterium-labeled version of Levofloxacin. Levofloxacin is a synthetic fluoroquinolone that acts as an antibacterial agent by inhibiting the supercoiling activity of bacterial DNA gyrase, thereby halting DNA replication.
  • Deuterium labeled levofloxacin
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only