Learn More
Abcam Levoflo x acin, Fluoroquinolone antibiotic, 1G
Catalog No. 502706768
Encompass_Preferred
MW 361.4 Da, Purity >98%. Fluoroquinolone antibiotic. Effective against Gram-positive and Gram-negative bacteria. Pharmacologically active following oral administration. Crosses the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6768 Supplier Abcam Supplier No. AB1412451G
MW 361.4 Da, Purity >98%. Fluoroquinolone antibiotic. Effective against Gram-positive and Gram-negative bacteria. Pharmacologically active following oral administration. Crosses the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement