Learn More
Selleck Chemical LLC Levofloxacin hydrate S4604-20mg
SDP
Catalog No. 503110951
Encompass_Preferred
Levofloxacin (Levaquin Tavanic Quixin Iquix Cravit) is a broad-spectrum third-generation fluoroquinolone antibiotic and optically active L-isomer of ofloxacin with antibacterial activity It acts by inhibiting DNA gyrase (bacterial topoisomerase II)
Catalog No. 50-311-0951 Supplier Selleck Chemical LLC Supplier No. S460420MG
Levofloxacin (Levaquin Tavanic Quixin Iquix Cravit) is a broad-spectrum third-generation fluoroquinolone antibiotic and optically active L-isomer of ofloxacin with antibacterial activity It acts by inhibiting DNA gyrase (bacterial topoisomerase II)