Learn More
Medchemexpress LLC Levofloxacin hydrate | 138199-71-0 | 100.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0330A10MM/1ML
Encompass_Preferred
Levofloxacin hydrate, also known as Levofloxacin hemihydrate, is an orally active antibiotic effective against both Gram-positive and Gram-negative bacteria. It functions by inhibiting DNA gyrase and topoisomerase IV. This compound is utilized in research for chronic periodontitis, airway inflammation, and BK Viremia, and has demonstrated anti-orthopoxvirus activity.
  • Orally active antibiotic.
  • Active against both Gram-positive and Gram-negative bacteria.
  • Inhibits DNA gyrase and topoisomerase IV.
  • Applicable for research in chronic periodontitis, airway inflammation, and BK Viremia.
  • Shows anti-orthopoxvirus activity.
Catalog No. 50-004-06761