Learn More
Medchemexpress LLC Levofloxacin (hydrochloride) | 177325-13-2 | 99.0% | 5 G
SDP
Catalog No. 5000405472
Encompass_Preferred
Levofloxacin hydrochloride is an orally active antibiotic that inhibits DNA gyrase and topoisomerase IV activity, inducing apoptosis. It demonstrates broad-spectrum antibacterial activity against both Gram-positive and Gram-negative bacteria, and has shown anticancer activity against lung cancer. This compound also exhibits anti-acnegenic, anxiogenic, and analgesic effects, and shortens sleep duration in mice, making it valuable for research into infectious diseases and lung cancer.
  • Orally active antibiotic
  • Inhibits DNA gyrase and topoisomerase IV activity
  • Induces apoptosis
  • Active against Gram-positive and Gram-negative bacteria
  • Exhibits anticancer activity against lung cancer
  • Possesses anti-acnegenic, anxiogenic, and analgesic effects
  • Shortens sleep duration in mice
  • Used in infectious diseases and lung cancer research
Catalog No. 50-004-05472 Supplier Medchemexpress LLC Supplier No. HYB0330B5G
Levofloxacin hydrochloride is an orally active antibiotic that inhibits DNA gyrase and topoisomerase IV activity, inducing apoptosis. It demonstrates broad-spectrum antibacterial activity against both Gram-positive and Gram-negative bacteria, and has shown anticancer activity against lung cancer. This compound also exhibits anti-acnegenic, anxiogenic, and analgesic effects, and shortens sleep duration in mice, making it valuable for research into infectious diseases and lung cancer.
  • Orally active antibiotic
  • Inhibits DNA gyrase and topoisomerase IV activity
  • Induces apoptosis
  • Active against Gram-positive and Gram-negative bacteria
  • Exhibits anticancer activity against lung cancer
  • Possesses anti-acnegenic, anxiogenic, and analgesic effects
  • Shortens sleep duration in mice
  • Used in infectious diseases and lung cancer research