Learn More
Cayman Chemical Levofloxacin N-oxide, Purity: ≥95%, CAS: 117678-38-3, 10 mg
SDP
Catalog No. 502939369
Encompass_Preferred
An inactive metabolite of levofloxacin; a degradation product of levofloxacin that is formed through exposure to daylight or hydrogen peroxide
Catalog No. 50-293-9369 Supplier Cayman Chemical Supplier No. 2717710 MG
An inactive metabolite of levofloxacin; a degradation product of levofloxacin that is formed through exposure to daylight or hydrogen peroxide